Chlorobis[dicyclohexyl(phenyl)phosphino](o-tolyl)nickel(II)

Supplier: TCI America
Danger

C3325-100MG C3325-500MG
TCC3325-100MGEA 106.11 CAD
TCC3325-100MG TCC3325-500MG
Chlorobis[dicyclohexyl(phenyl)phosphino](o-tolyl)nickel(II)
Chlorobis[dicyclohexyl(phenyl)phosphino](o-tolyl)nickel(II)
Formula: C₄₃H₆₁ClNiP₂
Melting Pt: 150°C
MDL Number: MFCD27978415
CAS Number: 1419179-26-2

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Elemental analysis (Carbon) 67.00 to 74.00 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR