10-(1,1':3',1''-Terphenyl-5'-yl)anthracene-9-boronic acid ≥97.0% (by HPLC) (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 10-(m-Terphenyl-5'-yl)anthracene-9-boronic acid

T3280-1G T3280-200MG
76220-550EA 598.82 CAD
76220-550 76220-552
10-(1,1':3',1''-Terphenyl-5'-yl)anthracene-9-boronic acid ≥97.0% (by HPLC) (contains varying amounts of Anhydride)
10-(1,1':3',1''-Terphenyl-5'-yl)anthracene-9-boronic acid
Formula: C₃₂H₂₃BO₂
MW: 450.34 g/mol
CAS Number: 1415334-59-6

Order Now

Specification Test Results

Appearance Light yellow to Yellow powder to crystal
Purity (HPLC) min. 97.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR