N,N,N',N'-Tetraphenyl-1,4-phenylenediamine ≥98.0% (by GC)
Supplier: TCI America
T3309-1G
T3309-5G
TCT3309-1GEA
77.42
CAD
TCT3309-1G
TCT3309-5G
N,N,N',N'-Tetraphenyl-1,4-phenylenediamine ≥98.0% (by GC)
N,N,N',N'-Tetraphenyl-1,4-phenylenediamine
Formula:
C₃₀H₂₄N₂ |
MDL Number:
MFCD08276846 CAS Number: 14118-16-2 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 201.0 to 205.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...