(R)-9-(2-Hydroxypropyl)adenine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
(R)-6-Amino-9-(2-hydroxypropyl)purine, (R)-1-(6-Amino-9H-purin-9-yl)propan-2-ol
H1291-1G
H1291-5G
TCH1291-1GEA
42.66
CAD
TCH1291-1G
TCH1291-5G
(R)-9-(2-Hydroxypropyl)adenine ≥98.0% (by HPLC, titration analysis)
(R)-9-(2-Hydroxypropyl)adenine
Formula:
C₈H₁₁N₅O MW: 193.21 g/mol |
MDL Number:
MFCD07369451 CAS Number: 14047-28-0 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 198.0 to 202.0 deg-C |
Specific rotation [a]20/D | -40.0 to -45.0 deg (C=1,EtOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...