N,N'-Di(2-naphthyl)-N,N'-diphenyl-1,4-phenylenediamine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

D4929-1G D4929-5G
TCD4929-1GEA 158 CAD
TCD4929-1G TCD4929-5G
N,N'-Di(2-naphthyl)-N,N'-diphenyl-1,4-phenylenediamine ≥98.0% (by HPLC, total nitrogen)
N,N'-Di(2-naphthyl)-N,N'-diphenyl-1,4-phenylenediamine
Formula: C₃₈H₂₈N₂
MW: 512.66 g/mol
MDL Number: MFCD28291914
CAS Number: 139994-47-1

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 192.0 to 196.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR