7,7'-Bi[1,4]benzoxazino[2,3,4-kl]phenoxazine ≥98.0% (by HPLC)
Supplier: TCI America
B4908-1G
B4908-200MG
TCB4908-1GEA
1167.62
CAD
TCB4908-1G
TCB4908-200MG
7,7'-Bi[1,4]benzoxazino[2,3,4-kl]phenoxazine ≥98.0% (by HPLC)
7,7'-Bi[1,4]benzoxazino[2,3,4-kl]phenoxazine
Formula:
C₃₆H₂₀N₂O₄ Melting Pt: 376°C |
CAS Number:
1395881-55-6 |
Specification Test Results
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 4.80 to 5.50 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...