9-(4-Biphenylyl)-2-bromocarbazole ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

B5110-1G B5110-200MG
TCB5110-1GEA 298.62 CAD
TCB5110-1G TCB5110-200MG
9-(4-Biphenylyl)-2-bromocarbazole ≥98.0% (by HPLC, total nitrogen)
9-(4-Biphenylyl)-2-bromocarbazole
Formula: C₂₄H₁₆BrN
MDL Number: MFCD27979953
CAS Number: 1393835-87-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 173.0 to 177.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR