9-([1,1'-Biphenyl]-4-yl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole ≥97.0% (by HPLC, total nitrogen)
Supplier: TCI America
|
Warning
|
Synonyms:
2-[9-([1,1'-Biphenyl]-4-yl)-9H-carbazol-3-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 9-(4-Biphenylyl)carbazole-3-boronic acid Pinacol ester
B5091-1G
B5091-5G
TCB5091-1GEA
58.46
CAD
TCB5091-1G
TCB5091-5G
9-([1,1'-Biphenyl]-4-yl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole ≥97.0% (by HPLC, total nitrogen)
9-([1,1'-Biphenyl]-4-yl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
Formula:
C₃₀H₂₈BNO₂ MW: 445.37 g/mol |
MDL Number:
MFCD29089348 CAS Number: 1391729-66-0 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Melting point | 229.0 to 233.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...