9-Benzyl-2,7-dibromo-9H-carbazole ≥98.0% (by GC, total nitrogen)
Supplier: TCI America
B5462-1G
B5462-5G
TCB5462-1GEA
224.36
CAD
TCB5462-1G
TCB5462-5G
9-Benzyl-2,7-dibromo-9H-carbazole ≥98.0% (by GC, total nitrogen)
9-Benzyl-2,7-dibromo-9H-carbazole
Formula:
C₁₉H₁₃Br₂N |
MDL Number:
MFCD30469065 CAS Number: 1384281-49-5 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (with Total Nitrogen) | min. 95.0 % |
Melting point | 161.0 to 165.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...