N,N'-Diphenyl-N,N'-bis(p-tolyl)-1,4-phenylenediamine ≥97.0% (by HPLC)
Supplier: TCI America
D4905-1G
D4905-5G
TCD4905-1GEA
120.08
CAD
TCD4905-1G
TCD4905-5G
N,N'-Diphenyl-N,N'-bis(p-tolyl)-1,4-phenylenediamine ≥97.0% (by HPLC)
N,N'-Diphenyl-N,N'-bis(p-tolyl)-1,4-phenylenediamine
Formula:
C₃₂H₂₈N₂ |
MDL Number:
MFCD01310665 CAS Number: 138171-14-9 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 95.0 % |
Melting point | 191.0 to 195.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...