1,5-Dimethyl-2,4-bis[2-(trimethylsilyl)ethynyl]benzene ≥97.0% (by GC)

Supplier: TCI America

D5029-1G D5029-5G
76219-792EA 142.2 CAD
76219-792 76219-794
1,5-Dimethyl-2,4-bis[2-(trimethylsilyl)ethynyl]benzene ≥97.0% (by GC)
1,5-Dimethyl-2,4-bis[2-(trimethylsilyl)ethynyl]benzene
Formula: C₁₈H₂₆Si₂
MW: 298.58 g/mol
Boiling Pt: 140°C
CAS Number: 1379822-08-8

Order Now

Specification Test Results

Purity (GC) min. 97.0 %
Melting point 55.0 to 59.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR