N,N'-Bis(4-formylphenyl)-N,N'-diphenylbenzidine ≥98.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
4,4'-Bis[N-(4-formylphenyl)-N-phenylamino]biphenyl
B4793-1G
B4793-200MG
TCB4793-1GEA
328.96
CAD
TCB4793-1G
TCB4793-200MG
N,N'-Bis(4-formylphenyl)-N,N'-diphenylbenzidine ≥98.0% (by total nitrogen basis)
N,N'-Bis(4-formylphenyl)-N,N'-diphenylbenzidine
Formula:
C₃₈H₂₈N₂O₂ MW: 544.65 g/mol |
MDL Number:
MFCD03093284 CAS Number: 137911-28-5 |
Specification Test Results
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...