5-Pyrazolone ≥98.0% (by GC)

Supplier: TCI America
Warning

P2563-1G P2563-200MG
TCP2563-1GEA 146.94 CAD
TCP2563-1G TCP2563-200MG
5-Pyrazolone ≥98.0% (by GC)
5-Pyrazolone
Formula: C₃H₄N₂O
MDL Number: MFCD00127223
CAS Number: 137-44-0

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 160.0 to 164.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR