N-(4-Biphenylyl)-2-biphenylamine ≥98.0% (by GC)

Supplier: TCI America
Warning

B4866-1G B4866-5G
TCB4866-1GEA 115.66 CAD
TCB4866-1G TCB4866-5G
N-(4-Biphenylyl)-2-biphenylamine ≥98.0% (by GC)
N-(4-Biphenylyl)-2-biphenylamine
Formula: C₂₄H₁₉N
MDL Number: MFCD22988894
CAS Number: 1372775-52-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 109.0 to 113.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR