5-Bromo-2-(5-bromo-2-thienyl)pyridine 98,0 GC_ASSAY_METHOD

Supplier: TCI America
Warning

Synonyms: 5-Bromo-2-(5-bromothiophen-2-yl)pyridine, 2-Bromo-5-(5-bromo-2-pyridyl)thiophene

B4886-1G B4886-200MG
TCB4886-1GEA 799.48 CAD
TCB4886-1G TCB4886-200MG
5-Bromo-2-(5-bromo-2-thienyl)pyridine 98,0 GC_ASSAY_METHOD
5-Bromo-2-(5-bromo-2-thienyl)pyridine
Formula: C₉H₅Br₂NS
MW: 319.01 g/mol
MDL Number: MFCD29765747
CAS Number: 136902-53-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 104.0 to 108.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR