Ethyl 3-Oxo-3-(2-thienyl)propionate ≥98.0% (by GC)

Supplier: TCI America

E1073-1G E1073-200MG
TCE1073-1GEA 183.28 CAD
TCE1073-1G TCE1073-200MG
Ethyl 3-Oxo-3-(2-thienyl)propionate ≥98.0% (by GC)
Ethyl 3-Oxo-3-(2-thienyl)propionate
Formula: C₉H₁₀O₃S
Boiling Pt: 130°C
MDL Number: MFCD00542649
CAS Number: 13669-10-8

Order Now

Specification Test Results

Appearance White or Colorless to Light yellow to Light orange powder to lump to clear liquid
Purity (GC) min. 98.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR