Hydrocortisone 17-Butyrate ≥98.0% (by HPLC)
Supplier: TCI America
|
Warning
|
H1506-1G
H1506-200MG
TCH1506-1GEA
206.66
CAD
TCH1506-1G
TCH1506-200MG
Hydrocortisone 17-Butyrate ≥98.0% (by HPLC)
Hydrocortisone 17-Butyrate
Formula:
C₂₅H₃₆O₆ |
MDL Number:
MFCD00083364 CAS Number: 13609-67-1 EINECS: 237-093-4 Merck Index: 14,04787 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 208.0 to 212.0 deg-C |
Specific rotation [a]20/D | +48.0 to +52.0 deg (C=0.1,CHCl3) (calcd.on dried substance) |
Drying loss | max. 1.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...