Pneumocandin B0 ≥96.0% (by HPLC)
Supplier: TCI America
Synonyms:
Pneumocandin B0, (10R,12S)-N-((2R,6S,9S,11R,12R,14aS,15S,20S,23S,25aS)-20-((R)-3-Amino-1-hydroxy-3-oxopropyl)-23-((1S,2S)-1,2-dihydroxy-2-(4-hydroxyphenyl)ethyl)-2,11,12,15-tetrahydroxy-6-((R)-1-hydroxyethyl)-5,8,14,19,22,25-hexaoxotetracosahydro-1H-dipyrrolo[2,1-c:2',1'-l][1,4,7,10,13,16]hexaazacyclohenicosin-9-yl)-10,12-dimethyltetradecanamide
P2534-10MG
P2534-50MG
TCP2534-10MGEA
151.68
CAD
TCP2534-10MG
TCP2534-50MG
Pneumocandin B0 ≥96.0% (by HPLC)
Pneumocandin B0
Formula:
C₅₀H₈₀N₈O₁₇ Storage Temperature: Freezer |
MDL Number:
MFCD28168025 CAS Number: 135575-42-7 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Elemental analysis (Nitrogen) | 9.50 to 11.50 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...