3,3'-Dimethyltriphenylamine ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
N,N-Di-m-tolylaniline, Bis(3-methylphenyl)phenylamine, Phenyl(di-m-tolyl)amine
D4755-1G
D4755-5G
TCD4755-1GEA
135.88
CAD
TCD4755-1G
TCD4755-5G
3,3'-Dimethyltriphenylamine ≥98.0% (by GC)
3,3'-Dimethyltriphenylamine
Formula:
C₂₀H₁₉N MW: 273.38 g/mol |
MDL Number:
MFCD23115649 CAS Number: 13511-11-0 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 70.0 to 74.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...