(3-Chloropropyl)diethoxy(methyl)silane ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
C3092-25ML
C3092-5ML
TCC3092-25MLEA
131.14
CAD
TCC3092-25ML
TCC3092-5ML
(3-Chloropropyl)diethoxy(methyl)silane ≥98.0% (by GC)
(3-Chloropropyl)diethoxy(methyl)silane
Formula:
C₈H₁₉ClO₂Si Boiling Pt: 109°C |
MDL Number:
MFCD05663910 CAS Number: 13501-76-3 EINECS: 236-828-6 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...