2-[2-(2-Aminoethoxy)ethoxy]acetic acid ≥96.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-(2-(2-Aminoethoxy)ethoxy)acetic acid, H2N-PEG2-CH2COOH, 8-Amino-3,6-dioxaoctanoic acid, Amino-PEG2-acetic acid

A3120-1G A3120-250MG
TCA3120-1GEA 529.3 CAD
TCA3120-1G TCA3120-250MG
2-[2-(2-Aminoethoxy)ethoxy]acetic acid ≥96.0% (by titrimetric analysis)
2-[2-(2-Aminoethoxy)ethoxy]acetic acid
Formula: C₆H₁₃NO₄
Storage Temperature: Freezer
MDL Number: MFCD13185897
CAS Number: 134978-97-5

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 124.0 to 128.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR