1,1'-Dibutyl-3,3,3',3'-tetramethylindotricarbocyanine Hexafluorophosphate ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 1-Butyl-2-[7-(1-butyl-1,3-dihydro-3,3-dimethyl-2H-indol-2-ylidene)-1,3,5-heptatrien-1-yl]-3,3-dimethyl-3H-indolium Hexafluorophosphate

D5013-1G D5013-5G
76219-136EA 259.12 CAD
76219-136 76219-138
1,1'-Dibutyl-3,3,3',3'-tetramethylindotricarbocyanine Hexafluorophosphate ≥98.0% (by HPLC)
1,1'-Dibutyl-3,3,3',3'-tetramethylindotricarbocyanine Hexafluorophosphate
Formula: C₃₅H₄₅F₆N₂P
MW: 638.72 g/mol
CAS Number: 134339-08-5

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Lambda max. 743.0 to 748.0 nm (MeOH)
Absorbance (E1%1cm) min. 4200 (MeOH, 743 to 748nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR