N,N'-Di(4-biphenylyl)-N,N'-diphenylbenzidine ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
D4928-1G
D4928-5G
TCD4928-1GEA
279.34
CAD
TCD4928-1G
TCD4928-5G
N,N'-Di(4-biphenylyl)-N,N'-diphenylbenzidine ≥98.0% (by HPLC, total nitrogen)
N,N'-Di(4-biphenylyl)-N,N'-diphenylbenzidine
Formula:
C₄₈H₃₆N₂ |
CAS Number:
134008-76-7 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 211.0 to 215.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...