2-Chloro-N-(2-chloro-4-methyl-3-pyridyl)nicotinamide ≥98.0% (by GC)

Supplier: TCI America

C3241-25G C3241-5G
TCC3241-25GEA 372.88 CAD
TCC3241-25G TCC3241-5G
2-Chloro-N-(2-chloro-4-methyl-3-pyridyl)nicotinamide ≥98.0% (by GC)
2-Chloro-N-(2-chloro-4-methyl-3-pyridyl)nicotinamide
Formula: C₁₂H₉Cl₂N₃O
MDL Number: MFCD09031256
CAS Number: 133627-46-0

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 193.0 to 197.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR