2,7-Dibromo-9-phenyl-9H-fluoren-9-ol ≥98.0% (by GC)

Supplier: TCI America

D5448-25G D5448-5G
76219-996EA 240.16 CAD
76219-996 76219-998
2,7-Dibromo-9-phenyl-9H-fluoren-9-ol ≥98.0% (by GC)
2,7-Dibromo-9-phenyl-9H-fluoren-9-ol
Formula: C₁₉H₁₂Br₂O
MDL Number: MFCD09953789
CAS Number: 132717-37-4

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 164.0 to 168.0 deg-C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR