1,5-Dibromo-2,6-dihydroxynaphthalene ≥98.0% (by GC)

Supplier: TCI America

D4668-25G D4668-5G
TCD4668-25GEA 1192.9 CAD
TCD4668-25G TCD4668-5G
1,5-Dibromo-2,6-dihydroxynaphthalene ≥98.0% (by GC)
1,5-Dibromo-2,6-dihydroxynaphthalene
Formula: C₁₀H₆Br₂O₂
MDL Number: MFCD16251539
CAS Number: 132178-78-0

Order Now

Specification Test Results

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR