2-(3-Bromophenyl)triphenylene ≥98.0% (by GC)
Supplier: TCI America
B4746-1G
B4746-200MG
TCB4746-1GEA
294.2
CAD
TCB4746-1G
TCB4746-200MG
2-(3-Bromophenyl)triphenylene ≥98.0% (by GC)
2-(3-Bromophenyl)triphenylene
Formula:
C₂₄H₁₅Br |
MDL Number:
MFCD28384139 CAS Number: 1313514-53-2 |
Specification Test Results
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 147.0 to 151.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...