Triethoxy[5,5,6,6,7,7,7-heptafluoro-4,4-bis(trifluoromethyl)heptyl]silane ≥90.0% (by GC)

Supplier: TCI America
Warning

T3246-1G T3246-5G
TCT3246-1GEA 142.2 CAD
TCT3246-1G TCT3246-5G
Triethoxy[5,5,6,6,7,7,7-heptafluoro-4,4-bis(trifluoromethyl)heptyl]silane ≥90.0% (by GC)
Triethoxy[5,5,6,6,7,7,7-heptafluoro-4,4-bis(trifluoromethyl)heptyl]silane
Formula: C₁₅H₂₁F₁₃O₃Si
MW: 524.39 g/mol
Boiling Pt: 81°C
MDL Number: MFCD27918526
CAS Number: 130676-81-2

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR