2-Amyl-5-methylphenol ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 5-Methyl-2-pentylphenol, Amylmetacresol

A2710-1G A2710-5G
TCA2710-1GEA 186.76 CAD
TCA2710-1G TCA2710-5G
2-Amyl-5-methylphenol ≥98.0% (by GC)
2-Amyl-5-methylphenol
Formula: C₁₂H₁₈O
MW: 178.28 g/mol
Boiling Pt: 139°C
MDL Number: MFCD00127710
CAS Number: 1300-94-3
EINECS: 215-094-0
UN: 3145
ADR: 8 III

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR