2-(tert-Butyldimethylsilyl)-N,N-dimethyl-1H-imidazole-1-sulfonamide ≥97.0% (by GC)

Supplier: TCI America
Warning

B5658-1G B5658-5G
76218-872EA 85.32 CAD
76218-872 76218-874
2-(tert-Butyldimethylsilyl)-N,N-dimethyl-1H-imidazole-1-sulfonamide ≥97.0% (by GC)
2-(tert-Butyldimethylsilyl)-N,N-dimethyl-1H-imidazole-1-sulfonamide
Formula: C₁₁H₂₃N₃O₂SSi
MDL Number: MFCD12963638
CAS Number: 129378-52-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 62.0 to 66.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR