2,2',6,6'-Tetra-tert-butyl-4,4'-dihydroxybiphenyl ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
2,2',6,6'-Tetra-tert-butyl-[1,1'-biphenyl]-4,4'-diol
T1807-1G
T1807-200MG
76220-506EA
358.66
CAD
76220-506
76220-508
2,2',6,6'-Tetra-tert-butyl-4,4'-dihydroxybiphenyl ≥98.0% (by GC)
2,2',6,6'-Tetra-tert-butyl-4,4'-dihydroxybiphenyl
Formula:
C₂₈H₄₂O₂ MW: 410.64 g/mol |
MDL Number:
MFCD00069443 CAS Number: 128-38-1 EINECS: 204-883-5 |
Specification Test Results
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 184.0 to 187.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...