Ancymidol ≥98.0% (by GC)
Supplier: TCI America
A2877-100MG
A2877-25MG
TCA2877-100MGEA
253.94
CAD
TCA2877-100MG
TCA2877-25MG
Ancymidol ≥98.0% (by GC)
Ancymidol
Formula:
C₁₅H₁₆N₂O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD00072501 CAS Number: 12771-68-5 EINECS: 235-814-7 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 109.0 to 113.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...