Diethylene Glycol bis(2-propynyl) ether ≥98.0% (by GC)
Supplier: TCI America
D4581-25G
D4581-5G
TCD4581-25GEA
892.7
CAD
TCD4581-25G
TCD4581-5G
Diethylene Glycol bis(2-propynyl) ether ≥98.0% (by GC)
Diethylene Glycol bis(2-propynyl) ether
Formula:
C₁₀H₁₄O₃ Boiling Pt: 103°C Storage Temperature: Refrigerator |
MDL Number:
MFCD22201523 CAS Number: 126422-57-9 |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...