Atorvastatin Lactone ≥95.0% (by HPLC)

Supplier: TCI America

A3114-100MG A3114-25MG
76219-286EA 573.54 CAD
76219-286 76219-288
Atorvastatin Lactone ≥95.0% (by HPLC)
Atorvastatin Lactone
Formula: C₃₃H₃₃FN₂O₄
Melting Pt: 161°C
MDL Number: MFCD00899262
CAS Number: 125995-03-1
Merck Index: 14,00864

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 95.0 area%
Elemental analysis (Nitrogen) 4.70 to 5.70 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR