2-(9,9-Diphenyl-9H-fluoren-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
D5033-1G
D5033-200MG
76219-802EA
346.02
CAD
76219-802
76219-804
2-(9,9-Diphenyl-9H-fluoren-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by HPLC, titration analysis)
2-(9,9-Diphenyl-9H-fluoren-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula:
C₃₁H₂₉BO₂ |
MDL Number:
MFCD30187268 CAS Number: 1259280-37-9 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 168.0 to 172.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...