4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzylamine hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
T3448-1G
T3448-200MG
TCT3448-1GEA
1036.48
CAD
TCT3448-1G
TCT3448-200MG
4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzylamine hydrochloride ≥98.0% (by HPLC, titration analysis)
4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzylamine hydrochloride
Formula:
C₉H₈F₃N₃·HCl Storage Temperature: Refrigerator |
MDL Number:
MFCD29765752 CAS Number: 1258874-29-1 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...