9-Phenyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

P2364-1G P2364-200MG
TCP2364-1GEA 341.28 CAD
TCP2364-1G TCP2364-200MG
9-Phenyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole ≥98.0% (by GC)
9-Phenyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
Formula: C₂₄H₂₄BNO₂
MDL Number: MFCD28129441
CAS Number: 1246669-45-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 147.0 to 151.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR