2-Amino-N-[(1,1'-biphenyl)-4-yl]-N-(4-bromophenyl)-9,9-dimethylfluorene 98,0 HPLC, N
Supplier: TCI America
Synonyms:
N-([1,1'-Biphenyl]-4-yl)-N-(4-bromophenyl)-9,9-dimethyl-9H-fluoren-2-amine
A2858-1G
A2858-5G
TCA2858-1GEA
52.93
CAD
TCA2858-1G
TCA2858-5G
2-Amino-N-[(1,1'-biphenyl)-4-yl]-N-(4-bromophenyl)-9,9-dimethylfluorene 98,0 HPLC, N
2-Amino-N-[(1,1'-biphenyl)-4-yl]-N-(4-bromophenyl)-9,9-dimethylfluorene
Formula:
C₃₃H₂₆BrN MW: 516.48 g/mol |
MDL Number:
MFCD20040455 CAS Number: 1246562-40-2 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 159.0 to 163.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...