2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: (9H-Carbazol-2-yl)boronic acid Pinacol ester, 2-(9H-Carbazol-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

T3441-1G T3441-5G
76220-638EA 194.34 CAD
76220-638 76220-640
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole ≥98.0% (by GC)
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
Formula: C₁₈H₂₀BNO₂
MDL Number: MFCD07784368
CAS Number: 1242412-60-7

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 195.0 to 200.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR