[8-(Glycidyloxy)-n-octyl]trimethoxysilane ≥97.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Trimethoxy(8-(oxiran-2-ylmethoxy)octyl)silane

G0469-25G G0469-5G
76220-102EA 527.72 CAD
76220-102 76220-104
[8-(Glycidyloxy)-n-octyl]trimethoxysilane ≥97.0% (by GC)
[8-(Glycidyloxy)-n-octyl]trimethoxysilane
Formula: C₁₄H₃₀O₅Si
MW: 306.47 g/mol
MDL Number: MFCD29472534
CAS Number: 1239602-38-0

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR