4-Amino-5-chloro-2,3-dihydrobenzofuran-7-carboxylic acid ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

A2918-1G A2918-200MG
TCA2918-1GEA 462.78 CAD
TCA2918-1G TCA2918-200MG
4-Amino-5-chloro-2,3-dihydrobenzofuran-7-carboxylic acid ≥97.0% (by HPLC, titration analysis)
4-Amino-5-chloro-2,3-dihydrobenzofuran-7-carboxylic acid
Formula: C₉H₈ClNO₃
MDL Number: MFCD12923204
CAS Number: 123654-26-2

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR