[4-(Trifluoromethyl)phenyl](2,4,6-trimethylphenyl)iodonium trifluoromethanesulfonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

T3446-1G T3446-5G
76220-646EA 202.24 CAD
76220-646 76220-648
[4-(Trifluoromethyl)phenyl](2,4,6-trimethylphenyl)iodonium trifluoromethanesulfonate ≥98.0% (by titrimetric analysis)
[4-(Trifluoromethyl)phenyl](2,4,6-trimethylphenyl)iodonium trifluoromethanesulfonate
Formula: C₁₇H₁₅F₆IO₃S
MDL Number: MFCD27978254
CAS Number: 1232133-62-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Ion exchange titration) min. 98.0 %
Melting point 201.0 to 205.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR