2,5-Dibromo-N-(2-ethylhexyl)-3,4-thiophenedicarboximide ≥98.0% (by GC)

Supplier: TCI America

D5021-1G D5021-200MG
76219-784EA 592.5 CAD
76219-784 76219-786
2,5-Dibromo-N-(2-ethylhexyl)-3,4-thiophenedicarboximide ≥98.0% (by GC)
2,5-Dibromo-N-(2-ethylhexyl)-3,4-thiophenedicarboximide
Formula: C₁₄H₁₇Br₂NO₂S
MDL Number: MFCD23703120
CAS Number: 1231160-83-0

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 95.0 %
Melting point 110.0 to 114.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR