2-Iodotriphenylene ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
I0978-1G
I0978-200MG
TCI0978-1GEA
680.98
CAD
TCI0978-1G
TCI0978-200MG
2-Iodotriphenylene ≥98.0% (by GC)
2-Iodotriphenylene
Formula:
C₁₈H₁₁I |
CAS Number:
1228778-59-3 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 155.0 to 159.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...