N,N-Bis(4-biphenylyl)aniline ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

B5086-1G B5086-5G
TCB5086-1GEA 191.18 CAD
TCB5086-1G TCB5086-5G
N,N-Bis(4-biphenylyl)aniline ≥98.0% (by HPLC, total nitrogen)
N,N-Bis(4-biphenylyl)aniline
Formula: C₃₀H₂₃N
MDL Number: MFCD20483034
CAS Number: 122215-84-3

Order Now

Specification Test Results

Appearance White to Almost white clear liquid
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 169.0 to 173.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR