9-([1,1'-Biphenyl]-3-yl)-9H-carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

B5591-1G B5591-5G
TCB5591-1GEA 99.54 CAD
TCB5591-1G TCB5591-5G
9-([1,1'-Biphenyl]-3-yl)-9H-carbazole ≥98.0% (by GC)
9-([1,1'-Biphenyl]-3-yl)-9H-carbazole
Formula: C₂₄H₁₇N
MW: 319.41 g/mol
MDL Number: MFCD28167065
CAS Number: 1221237-87-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 126.0 to 130.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR