2-(2-Propynyloxy)ethylamine ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

P2166-1G P2166-5G
TCP2166-1GEA 576.86 CAD
TCP2166-1G TCP2166-5G
2-(2-Propynyloxy)ethylamine ≥98.0% (by titrimetric analysis)
2-(2-Propynyloxy)ethylamine
Formula: C₅H₉NO
Storage Temperature: Refrigerator
MDL Number: MFCD16713446
CAS Number: 122116-12-5
UN: 2734
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR