9-Phenyl-3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-9H-carbazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

P2561-1G P2561-5G
TCP2561-1GEA 192.76 CAD
TCP2561-1G TCP2561-5G
9-Phenyl-3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-9H-carbazole ≥98.0% (by HPLC, titration analysis)
9-Phenyl-3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-9H-carbazole
Formula: C₃₀H₂₈BNO₂
MDL Number: MFCD30187279
CAS Number: 1219956-30-5

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 144.0 to 148.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR