9-(2'-Bromo-4-biphenylyl)carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

B5312-1G B5312-200MG
TCB5312-1GEA 316 CAD
TCB5312-1G TCB5312-200MG
9-(2'-Bromo-4-biphenylyl)carbazole ≥98.0% (by GC)
9-(2'-Bromo-4-biphenylyl)carbazole
Formula: C₂₄H₁₆BrN
MDL Number: MFCD31618099
CAS Number: 1215228-57-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 104.0 to 108.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR