1,3-Diaminopropane dihydroiodide ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
1,3-Propanediamine dihydroiodide, Trimethylenediamine dihydroiodide
D5091-1G
D5091-5G
76219-146EA
178.54
CAD
76219-146
76219-148
1,3-Diaminopropane dihydroiodide ≥98.0% (by total nitrogen and titration analysis)
1,3-Diaminopropane dihydroiodide
Formula:
C₃H₁₀N₂·₂HI MW: 329.95 g/mol |
CAS Number:
120675-53-8 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...